missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Color | White |
| Quantity | 250 mg |
| Formula Weight | 366.38 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |