CAS RN 942518-20-9
IUPAC Name:
4-azido-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms:
4-azido-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol):
366.38
Molecular Formula:
C19H18N4O4
InChi Key:
CLEZARXVEABQBI-UHFFFAOYNA-N
SMILES:
OC(=O)C(CCN=[N+]=[N-])NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine 98.0+%, TCI America™
CAS: 942518-20-9 Molecular Formula: C19H18N4O4 Molecular Weight (g/mol): 366.377 MDL Number: MFCD11052920 InChI Key: CLEZARXVEABQBI-KRWDZBQOSA-N Synonym: 4-Azido-N-Fmoc-L-homoalanine PubChem CID: 25146060 IUPAC Name: (2S)-4-azido-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid SMILES: C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN=[N+]=[N-])C(=O)O