missing translation for 'onlineSavingsMsg'
Learn More

4-Aminosalicylic Acid 98.0+%, TCI America™

Catalog No. p-7116891
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A042025G 25 g
A0420100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A042025G Supplier TCI America Supplier No. A042025G
Only null left

Chemical Identifiers

CAS 65-49-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007789
InChI Key WUBBRNOQWQTFEX-UHFFFAOYSA-N
Synonym 4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil
PubChem CID 4649
ChEBI CHEBI:27565
IUPAC Name 4-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)O)C(=O)O

Specifications

Melting Point 151°C
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminosalicylic Acid
EINECSNumber (3)-3039
RTECSNumber VO1225000
TSCA Yes