missing translation for 'onlineSavingsMsg'
Learn More

4-Aminosalicylic acid, 98+%

Catalog No. p-7026940
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2328922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2328922 Supplier Thermo Scientific Chemicals Supplier No. B2328922
Only null left

CAS: 65-49-6 | C7H7NO3 | 153.137 g/mol

Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.

Solubility
Soluble in water (2 g/L) at 20°C, dilute sodium hydroxide, dilute nitric acid, ether (slightly), methanol, acetone, and ethanol. Virtually insoluble in benzene, chloroform, and carbon tetrachloride.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 65-49-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007789
InChI Key WUBBRNOQWQTFEX-UHFFFAOYSA-N
Synonym 4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil
PubChem CID 4649
ChEBI CHEBI:27565
IUPAC Name 4-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)O)C(=O)O

Specifications

Melting Point ∼135°C to 145°C (decomposition)
Odor Odorless
Quantity 100 g
Beilstein 473071
Merck Index 14,477
Solubility Information Soluble in water (2g/L) at 20°C,dilute sodium hydroxide,dilute nitric acid,ether (slightly),methanol,acetone,and ethanol. Virtually insoluble in benzene,chloroform,and carbon tetrachloride.
Formula Weight 153.14
Percent Purity ≥98%
Chemical Name or Material 4-Aminosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-613-5
RTECSNumber VO1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only