missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminophenol Hydrochloride 98.0+%, TCI America™
Supplier: TCI America A0392100G
Specifications
| 4-Aminophenol Hydrochloride | |
| 306°C | |
| C6H8ClNO | |
| 100 g | |
| RVGOBWDGAVAVPJ-UHFFFAOYSA-N | |
| 4-aminophenol;hydrochloride | |
| 5828 | |
| ≥98.0% (HPLC,N) |
| 51-78-5 | |
| White-Yellow | |
| MFCD00012996 | |
| 4-aminophenol hydrochloride, futramine p, p-aminophenol hydrochloride, 4-hydroxyanilinium chloride, fouramine cp, fourrine p, durafur brown r, pelagol cp, peltol p, pelagol grey cp | |
| C1=CC(=CC=C1N)O.Cl | |
| 145.586 | |
| 145.59 | |
| Crystalline Powder |
Chemical Identifiers
| 51-78-5 | |
| 145.586 | |
| RVGOBWDGAVAVPJ-UHFFFAOYSA-N | |
| 5828 | |
| C1=CC(=CC=C1N)O.Cl |
| C6H8ClNO | |
| MFCD00012996 | |
| 4-aminophenol hydrochloride, futramine p, p-aminophenol hydrochloride, 4-hydroxyanilinium chloride, fouramine cp, fourrine p, durafur brown r, pelagol cp, peltol p, pelagol grey cp | |
| 4-aminophenol;hydrochloride |
Safety and Handling
EINECSNumber : (3)-0675
RTECSNumber : SJ6070000
TSCA : Yes