missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7116872
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A039225G 25 g
A0392100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A039225G Supplier TCI America Supplier No. A039225G
Only null left

Chemical Identifiers

CAS 51-78-5
Molecular Formula C6H8ClNO
Molecular Weight (g/mol) 145.586
MDL Number MFCD00012996
InChI Key RVGOBWDGAVAVPJ-UHFFFAOYSA-N
Synonym 4-aminophenol hydrochloride, futramine p, p-aminophenol hydrochloride, 4-hydroxyanilinium chloride, fouramine cp, fourrine p, durafur brown r, pelagol cp, peltol p, pelagol grey cp
PubChem CID 5828
IUPAC Name 4-aminophenol;hydrochloride
SMILES C1=CC(=CC=C1N)O.Cl

Specifications

Melting Point 306°C
Color White-Yellow
Quantity 25 g
Formula Weight 145.59
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenol Hydrochloride
EINECSNumber (3)-0675
RTECSNumber SJ6070000
TSCA Yes