Learn More
4-Aminophenol, 98%
CAS: 123-30-8 | C6H7NO | 109.128 g/mol
Supplier: Thermo Scientific Chemicals A135810I
Description
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.
Solubility
Soluble in acetonitrile, ethyl acetate, acetone, hot water, dimethylsulfoxide and alkalies. Slightly soluble in toluene, diethyl ether, ethanol, trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.
Notes
Incompatible with acids, chloroformates and strong oxidizing agents.
Specifications
| 4-Aminophenol | |
| 186°C to 190°C | |
| 284°C (decomposition) | |
| 98% | |
| C6H7NO | |
| 5000 g | |
| 385836 | |
| 14,462 | |
| Soluble in acetonitrile,ethyl acetate,acetone,hot water,dimethylsulfoxide and alkalies. Slightly soluble in toluene,diethyl ether,ethanol,trifluoroacetic acid and cold water. Insoluble in benzene and chloroform. | |
| C1=CC(=CC=C1N)O | |
| 109.128 | |
| CHEBI:17602 | |
| 98% |
| 123-30-8 | |
| 1.29 | |
| 195°C (383°F) | |
| Phenol-like | |
| MFCD00007869 | |
| UN2512 | |
| Air and light sensitive | |
| p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p | |
| PLIKAWJENQZMHA-UHFFFAOYSA-N | |
| 4-aminophenol | |
| 403 | |
| 109.13 |
Chemical Identifiers
| 123-30-8 | |
| 109.128 | |
| PLIKAWJENQZMHA-UHFFFAOYSA-N | |
| 403 | |
| 4-aminophenol |
| C6H7NO | |
| MFCD00007869 | |
| p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p | |
| CHEBI:17602 | |
| C1=CC(=CC=C1N)O |
Safety and Handling
GHS H Statement
H341-H302-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P280-P301+P312-P304+P340-P308+P313-P312-P330-P501c
H302+H332-H341
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-)
EINECSNumber : 204-616-2
RTECSNumber : SJ5075000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only