missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzonitrile Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117559
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A16451G 1 g
A16455G 5 g
2 options

Catalog No. A16451G

Supplier: TCI America A16451G

Only null left

Chemical Identifiers

CAS 15996-76-6
Molecular Formula C8H9ClN2
Molecular Weight (g/mol) 168.624
MDL Number MFCD01861472
InChI Key QREZLLYPLRPULF-UHFFFAOYSA-N
Synonym 4-aminomethyl benzonitrile hydrochloride, 4-cyanobenzylamine hcl, 4-cyanobenzylamine hydrochloride, 4-aminomethyl benzonitrile hcl, 4-aminomethyl-benzonitrile hydrochloride, benzonitrile, 4-aminomethyl-, monohydrochloride, p-aminomethyl benzonitrile hydrochloride, pubchem2057, acmc-209dkg, 4-aminomethylbenzonitrile hcl
PubChem CID 9942250
IUPAC Name 4-(aminomethyl)benzonitrile;hydrochloride
SMILES C1=CC(=CC=C1CN)C#N.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 168.62
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Aminomethyl)benzonitrile Hydrochloride