accessibility menu, dialog, popup

UserName

CAS RN 15996-76-6

IUPAC Name: hydrogen 4-(aminomethyl)benzonitrile chloride
Synonyms: hydrogen 4-(aminomethyl)benzonitrile chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C8H9ClN2
InChi Key: QREZLLYPLRPULF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)