missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzonitrile hydrochloride, 97%

Catalog No. p-7029950
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2747306 5 g
AAH2747314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2747306 Supplier Thermo Scientific Chemicals Supplier No. H2747306

CAS: 15996-76-6 | C8H9ClN2 | 168.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15996-76-6
Molecular Formula C8H9ClN2
Molecular Weight (g/mol) 168.624
MDL Number MFCD01861472
InChI Key QREZLLYPLRPULF-UHFFFAOYSA-N
Synonym 4-aminomethyl benzonitrile hydrochloride, 4-cyanobenzylamine hcl, 4-cyanobenzylamine hydrochloride, 4-aminomethyl benzonitrile hcl, 4-aminomethyl-benzonitrile hydrochloride, benzonitrile, 4-aminomethyl-, monohydrochloride, p-aminomethyl benzonitrile hydrochloride, pubchem2057, acmc-209dkg, 4-aminomethylbenzonitrile hcl
PubChem CID 9942250
IUPAC Name 4-(aminomethyl)benzonitrile;hydrochloride
SMILES C1=CC(=CC=C1CN)C#N.Cl

Specifications

Melting Point 279°C to 284°C
Quantity 5 g
Formula Weight 168.62
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)benzonitrile hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only