missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzeneboronic acid hydrochloride, 96%

Catalog No. p-7035141
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5285503 1 g
AAH5285506 5 g
2 options

Catalog No. AAH5285503

Supplier: Thermo Scientific Chemicals H5285503

Only null left

CAS: 75705-21-4 | C7H11BClNO2 | 187.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 75705-21-4
Molecular Formula C7H11BClNO2
Molecular Weight (g/mol) 187.43
MDL Number MFCD01632199
InChI Key HUZNRXFJHYNUMV-UHFFFAOYSA-N
Synonym 4-aminomethylphenylboronic acid hydrochloride, 4-aminomethyl phenylboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hcl, 4-aminomethyl benzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid hcl, 4-aminomethylphenyl boronic acid hydrochloride, 4-aminomethyl phenyl boronic acid hydrochloride, 4-aminomethylbenzeneboronic acid hydrochloride, 4-aminomethylphenylboronic acid, hydrochloride
PubChem CID 2734311
IUPAC Name [4-(aminomethyl)phenyl]boronic acid;hydrochloride
SMILES Cl.NCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 240°C to 250°C
Quantity 1 g
Formula Weight 187.43
Percent Purity 96%
Chemical Name or Material 4-(Aminomethyl)benzeneboronic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only