missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric Acid 99.0+%, TCI America™

Catalog No. A031310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A031310G 10 g
A031325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A031310G Supplier TCI America Supplier No. A031310G

Chemical Identifiers

CAS 61-78-9
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00007890
InChI Key HSMNQINEKMPTIC-UHFFFAOYSA-N
Synonym 4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
PubChem CID 2148
ChEBI CHEBI:104011
IUPAC Name 2-[(4-aminobenzoyl)amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)O)N

Specifications

Melting Point 197°C
Quantity 10 g
Formula Weight 194.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminohippuric Acid
EINECSNumber (9)-0111
TSCA Yes