accessibility menu, dialog, popup

UserName

H2NONHOHO

CAS RN 61-78-9

IUPAC Name: 2-[(4-aminophenyl)formamido]acetic acid
Synonyms: aminohippurate sodium para-aminohippurate pah (amino acid) nefrotest aminohippurate sodium preparation nephrotest PAH pah - para-aminohippurate 氨马尿酸 Paha
Molecular Weight (g/mol): 194.19
Molecular Formula: C9H10N2O3
InChi Key: HSMNQINEKMPTIC-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(=O)NCC(O)=O
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)