missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric acid, 99%

Catalog No. AC153800100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153800100 10 g Glass Bottle
AC153800250 25 g Plastic bottle
AC153801000 100 g Plastic bottle
AC153805000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC153800100 Supplier Thermo Scientific Chemicals Supplier No. 153800100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 61-78-9 | C9H10N2O3 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61-78-9
Molecular Weight (g/mol) 194.19
MDL Number MFCD00007890
InChI Key HSMNQINEKMPTIC-UHFFFAOYSA-N
Synonym 4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
PubChem CID 2148
ChEBI CHEBI:104011
IUPAC Name 2-[(4-aminobenzoyl)amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C9H10N2O3
Linear Formula H2NC6H4CONHCH2CO2H
Quantity 10 g
Beilstein 14, II, 258
Merck Index 15, 439
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, benzene, aceton and most, organic solvents, pract. insoluble in ccl4, very soluble in dilute hydrochloric acid and alkal
Formula Weight 194.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminohippuric acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-518-9

RUO – Research Use Only