missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminobutyric Acid, Reagent, 98%, Spectrum™ Chemical
AM151, 56-12-2, C4H9NO2
$196.82 - $18096.30
Chemical Identifiers
| CAS | 56-12-2 |
|---|---|
| Molecular Formula | C4H9NO2 |
| Molecular Weight (g/mol) | 103.12 |
| InChI Key | BTCSSZJGUNDROE-UHFFFAOYSA-N |
| IUPAC Name | 4-aminobutanoic acid |
| SMILES | NCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600133
|
Spectrum Chemical Mfg Cor
AM15125GM |
25 g | Poly Bottle |
Each for $196.82
|
|
||||
|
18600134
|
Spectrum Chemical Mfg Cor
AM151100GM |
100 g | Poly Bottle |
Each for $326.45
|
|
||||
|
18600135
|
Spectrum Chemical Mfg Cor
AM151500GM |
500 g | Poly Bottle |
Each for $1,157.70
|
|
||||
|
18600137
|
Spectrum Chemical Mfg Cor
AM15112KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18600136
|
Spectrum Chemical Mfg Cor
AM15125KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||