missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminobutyric Acid, Reagent, 98%, Spectrum™ Chemical
Description
Spectrum™ Chemical 4-Aminobutyric Acid, Reagent belongs to the transaminases family which transfer nitrogenous groups. It is an enzyme which plays a part in 5 metabolic pathways: glutamate metabolism, beta-alanine metabolism, alanine and aspartate metabolism, butanoate metabolism, and propionate metabolism. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.33% |
| Loss on Drying | 0.5% |
| Packaging | Poly Pail |
| Quantity | 12 kg |
| Percent Purity | 98% |
| Grade | Reagent |