missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric Acid, Reagent, 98%, Spectrum™ Chemical

Catalog No. 18600133
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600133 25 g Poly Bottle
18600134 100 g Poly Bottle
18600135 500 g Poly Bottle
18600137 12 kg Poly Pail
18600136 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600133 Supplier Spectrum Chemical Mfg Cor Supplier No. AM15125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

AM151, 56-12-2, C4H9NO2

Spectrum™ Chemical 4-Aminobutyric Acid, Reagent belongs to the transaminases family which transfer nitrogenous groups. It is an enzyme which plays a part in 5 metabolic pathways: glutamate metabolism, beta-alanine metabolism, alanine and aspartate metabolism, butanoate metabolism, and propionate metabolism. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
IUPAC Name 4-aminobutanoic acid
SMILES NCCCC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.33%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98%
Grade Reagent