missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid sodium salt, 98%

Pharmaceutical intermediate | CAS: 555-06-6 | C7H6NNaO2 | 159.12 g/mol

Supplier:  Thermo Scientific Chemicals J6342822

 View more versions of this product

Catalog No. AAJ6342822


Only null left
Explore available promotions

Description

Description

4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

Solubility
Soluble in water (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-Aminobenzoic acid sodium salt
>300°C
C7H6NNaO2
100 g
Soluble in water (50mg/ml).
C1=CC(=CC=C1C(=O)[O-])N.[Na+]
159.12
159.12
Powder
555-06-6
Yellow
MFCD00064395
sodium 4-aminobenzoate, sodium p-aminobenzoate, 4-aminobenzoic acid sodium salt, sodium aminobenzoate, aminobenzoate sodium, antergyl, pabavit, benzoic acid, 4-amino-, monosodium salt, monosodium 4-aminobenzoate, unii-75ui7quz5j
XETSAYZRDCRPJY-UHFFFAOYSA-M
sodium;4-aminobenzoate
517441
98%

Chemical Identifiers

555-06-6
159.12
XETSAYZRDCRPJY-UHFFFAOYSA-M
517441
C1=CC(=CC=C1C(=O)[O-])N.[Na+]
C7H6NNaO2
MFCD00064395
sodium 4-aminobenzoate, sodium p-aminobenzoate, 4-aminobenzoic acid sodium salt, sodium aminobenzoate, aminobenzoate sodium, antergyl, pabavit, benzoic acid, 4-amino-, monosodium salt, monosodium 4-aminobenzoate, unii-75ui7quz5j
sodium;4-aminobenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 209-080-3

RTECSNumber : DG2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only