missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid sodium salt, 98%

Catalog No. AAJ6342814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6342814 25 g
AAJ6342822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6342814 Supplier Thermo Scientific Chemicals Supplier No. J6342814

Pharmaceutical intermediate | CAS: 555-06-6 | C7H6NNaO2 | 159.12 g/mol

4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

Solubility
Soluble in water (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 555-06-6
Molecular Formula C7H6NNaO2
Molecular Weight (g/mol) 159.12
MDL Number MFCD00064395
InChI Key XETSAYZRDCRPJY-UHFFFAOYSA-M
Synonym sodium 4-aminobenzoate, sodium p-aminobenzoate, 4-aminobenzoic acid sodium salt, sodium aminobenzoate, aminobenzoate sodium, antergyl, pabavit, benzoic acid, 4-amino-, monosodium salt, monosodium 4-aminobenzoate, unii-75ui7quz5j
PubChem CID 517441
IUPAC Name sodium;4-aminobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])N.[Na+]

Specifications

Melting Point >300°C
Color Yellow
Quantity 25 g
Solubility Information Soluble in water (50mg/ml).
Formula Weight 159.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminobenzoic acid sodium salt
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 209-080-3
RTECSNumber DG2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only