missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzenethiol 98.0+%, TCI America™

Supplier:  TCI America A06235G

 View more versions of this product

Catalog No. A06235G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Aminobenzenethiol
Yellow
C6H6NS
5 g
4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan
NC1=CC=C([S-])C=C1
124.18
125.19
Crystalline Lumps
1193-02-8
142°C
MFCD00007880
3263
WCDSVWRUXWCYFN-UHFFFAOYSA-M
(4-aminophenyl)sulfanide
14510
≥98.0% (GC)

Chemical Identifiers

1193-02-8
124.18
WCDSVWRUXWCYFN-UHFFFAOYSA-M
14510
NC1=CC=C([S-])C=C1
C6H6NS
MFCD00007880
4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan
(4-aminophenyl)sulfanide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DC0602500

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions