missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzenethiol 98.0+%, TCI America™

Catalog No. p-7116971
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06235G 5 g
A062325G 25 g
A0623100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A06235G Supplier TCI America Supplier No. A06235G
Only null left

Chemical Identifiers

CAS 1193-02-8
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007880
InChI Key WCDSVWRUXWCYFN-UHFFFAOYSA-M
Synonym 4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan
PubChem CID 14510
IUPAC Name (4-aminophenyl)sulfanide
SMILES NC1=CC=C([S-])C=C1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 3263
Formula Weight 125.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Aminobenzenethiol
RTECSNumber DC0602500
TSCA No
Recommended Storage Refrigerator