missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-p-terphenyl 97.0+%, TCI America™

Catalog No. A13405G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13405G 5 g
A13401G 1 g
A134025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A13405G Supplier TCI America Supplier No. A13405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7293-45-0
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
MDL Number MFCD00191302
InChI Key ATGIXVUZFPZOHP-UHFFFAOYSA-N
PubChem CID 350072
IUPAC Name 4-(4-phenylphenyl)aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N

Specifications

Melting Point 201°C
Quantity 5 g
Formula Weight 245.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-p-terphenyl