accessibility menu, dialog, popup

UserName

CAS RN 7293-45-0

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}aniline
Synonyms: 4-{[1,1'-biphenyl]-4-yl}aniline
Molecular Weight (g/mol): 245.33
Molecular Formula: C18H15N
InChi Key: ATGIXVUZFPZOHP-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results