missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N-methylbenzamide 98.0+%, TCI America™

Catalog No. A20641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20641G 1 g
A20645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20641G Supplier TCI America Supplier No. A20641G
Only null left

Chemical Identifiers

CAS 6274-22-2
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
InChI Key XAGFYNSCWICYPA-UHFFFAOYSA-N
PubChem CID 235516
IUPAC Name 4-amino-N-methylbenzamide
SMILES CNC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 181°C
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-N-methylbenzamide