accessibility menu, dialog, popup

UserName

CH3HNONH2

CAS RN 6274-22-2

IUPAC Name: 4-amino-N-methylbenzamide
Synonyms: 4-amino-N-methylbenzamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: XAGFYNSCWICYPA-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results