missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-m-cresol 98.0+%, TCI America™

Catalog No. A028825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A028825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A028825G Supplier TCI America Supplier No. A028825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2835-99-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007871
InChI Key QGNGOGOOPUYKMC-UHFFFAOYSA-N
Synonym 4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
PubChem CID 17819
ChEBI CHEBI:55546
IUPAC Name 4-amino-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)N

Specifications

Melting Point 175°C
Quantity 25 g
UN Number 3077
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-m-cresol
EINECSNumber (3)-0707
RTECSNumber GO6826000
TSCA Yes