accessibility menu, dialog, popup

UserName

CH3OHH2N

CAS RN 2835-99-6

IUPAC Name: 4-amino-3-methylphenol
Synonyms: 4-amino-m-cresol 4-hydroxy-6'-methylalanine 4-OH-6'-methylalanine 4-amino-M-cresol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: QGNGOGOOPUYKMC-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)