missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-m-cresol, 99+%

Catalog No. p-55840
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163200500 50 g Glass bottle
AC163202500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163200500 Supplier Thermo Scientific Chemicals Supplier No. 163200500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2835-99-6 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2835-99-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007871
InChI Key QGNGOGOOPUYKMC-UHFFFAOYSA-N
Synonym 4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
PubChem CID 17819
ChEBI CHEBI:55546
IUPAC Name 4-amino-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 180°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)OH
Quantity 50 g
Beilstein 13, 593
Solubility Information Solubility in water: insoluble
Formula Weight 123.15
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 4-Amino-m-cresol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-621-2
RTECSNumber GO6826000
TSCA TSCA

RUO – Research Use Only