missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%

CAS: 121-30-2 | C6H8ClN3O4S2 | 285.73 g/mol

$58.87 - $58.87

Chemical Identifiers

CAS 121-30-2
Molecular Formula C6H8ClN3O4S2
Molecular Weight (g/mol) 285.73
InChI Key IHJCXVZDYSXXFT-UHFFFAOYSA-N
Synonym 4-amino-6-chloro-1,3-benzenedisulfonamide, chloraminophenamide, salmid, idorese, chloroaminophenamide, 1,3-benzenedisulfonamide, 4-amino-6-chloro, 5-chloro-2,4-disulfamylaniline, 4-amino-6-chloro-m-benzenedisulfonamide, 3-chloro-4,6-disulfamoylaniline, 5-chloro-2,4-disulfamoylaniline
PubChem CID 67136
ChEBI CHEBI:3602
IUPAC Name 4-amino-6-chlorobenzene-1,3-disulfonamide
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)N)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC251850050
SDS View Documents
Thermo Scientific Chemicals
251850050
5 g Glass bottle
Each for $58.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-30-2
285.73
4-amino-6-chloro-1,3-benzenedisulfonamide, chloraminophenamide, salmid, idorese, chloroaminophenamide, 1,3-benzenedisulfonamide, 4-amino-6-chloro, 5-chloro-2,4-disulfamylaniline, 4-amino-6-chloro-m-benzenedisulfonamide, 3-chloro-4,6-disulfamoylaniline, 5-chloro-2,4-disulfamoylaniline
CHEBI:3602
C1=C(C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)N)N
C6H8ClN3O4S2
IHJCXVZDYSXXFT-UHFFFAOYSA-N
67136
4-amino-6-chlorobenzene-1,3-disulfonamide

Specifications

121-30-2
100.0
White
97.5% min. (HPLC)
C6H8ClN3O4S2
15,2076
IHJCXVZDYSXXFT-UHFFFAOYSA-N
4-amino-6-chlorobenzene-1,3-disulfonamide
67136
285.73
Powder
97.5
260.0°C to 263.0°C
Authentic
Glass bottle
5 g
4-amino-6-chloro-1,3-benzenedisulfonamide, chloraminophenamide, salmid, idorese, chloroaminophenamide, 1,3-benzenedisulfonamide, 4-amino-6-chloro, 5-chloro-2,4-disulfamylaniline, 4-amino-6-chloro-m-benzenedisulfonamide, 3-chloro-4,6-disulfamoylaniline, 5-chloro-2,4-disulfamoylaniline
C1=C(C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)N)N
285.73
CHEBI:3602
98%
4-Amino-6-chloro-1, 3-benzenedisulfonamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
Rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Get medical attention/advice if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-463-1

SDS
Documents

Documents

RUO – Research Use Only