missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. A26155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26155G 5 g
A26151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26155G Supplier TCI America Supplier No. A26155G

Chemical Identifiers

CAS 71675-87-1
Molecular Formula C10H13NO5S
Molecular Weight (g/mol) 259.276
MDL Number MFCD04973619
InChI Key OJVNCXHGGYYOPH-UHFFFAOYSA-N
PubChem CID 3018232
IUPAC Name 4-amino-5-ethylsulfonyl-2-methoxybenzoic acid
SMILES CCS(=O)(=O)C1=C(C=C(C(=C1)C(=O)O)OC)N

Specifications

Melting Point 150°C
Quantity 5 g
Formula Weight 259.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic Acid