accessibility menu, dialog, popup

UserName

H3CSOOOHOOCH3H2N

CAS RN 71675-87-1

IUPAC Name: 4-amino-5-(ethanesulfonyl)-2-methoxybenzoic acid
Synonyms: 4-amino-5-(ethanesulfonyl)-2-methoxybenzoic acid
Molecular Weight (g/mol): 259.28
Molecular Formula: C10H13NO5S
InChi Key: OJVNCXHGGYYOPH-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)C1=CC(C(O)=O)=C(OC)C=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results