missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-4'-nitrobiphenyl 96.0+%, TCI America™
$354.17 - $1206.05
Chemical Identifiers
| CAS | 1211-40-1 |
|---|---|
| Molecular Formula | C12H10N2O2 |
| Molecular Weight (g/mol) | 214.224 |
| MDL Number | MFCD00093527 |
| InChI Key | BOFVBIYTBGDQGY-UHFFFAOYSA-N |
| PubChem CID | 14593 |
| IUPAC Name | 4-(4-nitrophenyl)aniline |
| SMILES | C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])N |
Chemical Identifiers
| 1211-40-1 | |
| 214.224 | |
| BOFVBIYTBGDQGY-UHFFFAOYSA-N | |
| 4-(4-nitrophenyl)aniline |
| C12H10N2O2 | |
| MFCD00093527 | |
| 14593 | |
| C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])N |
Specifications
| 1211-40-1 | |
| C12H10N2O2 | |
| 1 g | |
| BOFVBIYTBGDQGY-UHFFFAOYSA-N | |
| 4-(4-nitrophenyl)aniline | |
| 14593 | |
| ≥96.0% (T) | |
| 4-Amino-4′-nitrobiphenyl |
| 204°C | |
| MFCD00093527 | |
| 4-Amino-4′C-nitrodiphenyl | |
| C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])N | |
| 214.224 | |
| 214.22 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DV0350000
TSCA : No