accessibility menu, dialog, popup

UserName

CAS RN 1211-40-1

NH2NOO

CAS RN 1211-40-1

IUPAC Name: 4'-nitro-[1,1'-biphenyl]-4-amine
Synonyms: 4'-nitro-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: BOFVBIYTBGDQGY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results