missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-(N,N-dimethylamino)stilbene 98.0+%, TCI America™

Catalog No. A02915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A02915G Supplier TCI America Supplier No. A02915G

Chemical Identifiers

CAS 22525-43-5
Molecular Formula C16H18N2
Molecular Weight (g/mol) 238.334
MDL Number MFCD00059189
InChI Key GCHSJPKVJSMRDX-ONEGZZNKSA-N
Synonym N,N-Dimethyl-4,4′C-stilbenediamine
PubChem CID 5376919
IUPAC Name 4-[(E)-2-[4-(dimethylamino)phenyl]ethenyl]aniline
SMILES CN(C)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)N

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 238.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-(N,N-dimethylamino)stilbene