accessibility menu, dialog, popup

UserName

H3CNCH3NH2(E)

CAS RN 22525-43-5

IUPAC Name: 4-[(1E)-2-(4-aminophenyl)ethenyl]-N,N-dimethylaniline
Synonyms: 4-[(1E)-2-(4-aminophenyl)ethenyl]-N,N-dimethylaniline
Molecular Weight (g/mol): 238.33
Molecular Formula: C16H18N2
InChi Key: GCHSJPKVJSMRDX-ONEGZZNKSA-N
SMILES: CN(C)C1=CC=C(\C=C\C2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results