missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-4'-methoxystilbene 97.0+%, TCI America™
$417.56 - $1427.87
Chemical Identifiers
| CAS | 7570-37-8 |
|---|---|
| Molecular Formula | C15H15NO |
| Molecular Weight (g/mol) | 225.291 |
| MDL Number | MFCD00059856 |
| InChI Key | FUKHOQPXEPBRFC-NSCUHMNNSA-N |
| PubChem CID | 5376491 |
| IUPAC Name | 4-[(E)-2-(4-methoxyphenyl)ethenyl]aniline |
| SMILES | COC1=CC=C(C=C1)C=CC2=CC=C(C=C2)N |
Chemical Identifiers
| 7570-37-8 | |
| 225.291 | |
| FUKHOQPXEPBRFC-NSCUHMNNSA-N | |
| 4-[(E)-2-(4-methoxyphenyl)ethenyl]aniline |
| C15H15NO | |
| MFCD00059856 | |
| 5376491 | |
| COC1=CC=C(C=C1)C=CC2=CC=C(C=C2)N |
Specifications
| 7570-37-8 | |
| Yellow | |
| MFCD00059856 | |
| 4′C-Methoxy-4-stilbenamine | |
| COC1=CC=C(C=C1)C=CC2=CC=C(C=C2)N | |
| 225.291 | |
| 225.29 | |
| Crystalline Powder |
| 173°C | |
| C15H15NO | |
| 1 g | |
| FUKHOQPXEPBRFC-NSCUHMNNSA-N | |
| 4-[(E)-2-(4-methoxyphenyl)ethenyl]aniline | |
| 5376491 | |
| ≥97.0% (GC,T) | |
| 4-Amino-4′-methoxystilbene |
Safety and Handling
RTECSNumber : CY0890140
TSCA : No