CAS RN 7570-37-8
CAS RN 7570-37-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Amino-4'-methoxystilbene 97.0+%, TCI America™
CAS: 7570-37-8 Molecular Formula: C15H15NO Molecular Weight (g/mol): 225.291 MDL Number: MFCD00059856 InChI Key: FUKHOQPXEPBRFC-NSCUHMNNSA-N PubChem CID: 5376491 IUPAC Name: 4-[(E)-2-(4-methoxyphenyl)ethenyl]aniline SMILES: COC1=CC=C(C=C1)C=CC2=CC=C(C=C2)N