missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-4'-cyanobiphenyl 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 185°C |
| Color | Yellow |
| Quantity | 5 g |
| Synonym | 4′-Aminobiphenyl-4-carbonitrile, 4-Amino-4′-cyanodiphenyl |
| Formula Weight | 194.24 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-4′-cyanobiphenyl |
Safety and Handling
| RTECSNumber | DV1763000 |
| TSCA | No |