accessibility menu, dialog, popup

UserName

CAS RN 4854-84-6

NH2N

CAS RN 4854-84-6

IUPAC Name: 4'-amino-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-amino-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: CPJQKNUJNWPAPH-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results