missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-cyanobiphenyl 95.0+%, TCI America™

Catalog No. A12491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12491G 1 g
A12495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12491G Supplier TCI America Supplier No. A12491G
Only null left

Chemical Identifiers

CAS 4854-84-6
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00191367
InChI Key CPJQKNUJNWPAPH-UHFFFAOYSA-N
PubChem CID 97193
IUPAC Name 4-(4-aminophenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)N

Specifications

Melting Point 185°C
Color Yellow
Quantity 1 g
Synonym 4′C-Aminobiphenyl-4-carbonitrile, 4-Amino-4′C-cyanodiphenyl
Formula Weight 194.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-cyanobiphenyl
RTECSNumber DV1763000
TSCA No