missing translation for 'onlineSavingsMsg'
Learn More

4'-Amino-3'-(trifluoromethyl)acetanilide 98.0+%, TCI America™

Catalog No. A18955G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18955G Supplier TCI America Supplier No. A18955G

Chemical Identifiers

CAS 1579-89-1
Molecular Formula C9H9F3N2O
Molecular Weight (g/mol) 218.18
MDL Number MFCD00270762
InChI Key QMCLCXKXDXBQLB-UHFFFAOYSA-N
Synonym 5-Acetamido-2-aminobenzotrifluoride, 4-Acetamido-2-(trifluoromethyl)aniline
PubChem CID 16227548
IUPAC Name N-[4-amino-3-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC(=C(N)C=C1)C(F)(F)F

Specifications

Melting Point 119°C
Quantity 5 g
Formula Weight 218.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Amino-3′-(trifluoromethyl)acetanilide