accessibility menu, dialog, popup

UserName

CH3ONHNH2FFF

CAS RN 1579-89-1

IUPAC Name: N-[4-amino-3-(trifluoromethyl)phenyl]acetamide
Synonyms: N-[4-amino-3-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 218.18
Molecular Formula: C9H9F3N2O
InChi Key: QMCLCXKXDXBQLB-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(=C(N)C=C1)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results