missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenol 97.0+%, TCI America™

Catalog No. p-7117429
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A142625G 25 g
A1426100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A142625G Supplier TCI America Supplier No. A142625G
Only null left

Chemical Identifiers

CAS 610-81-1
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00066310
InChI Key IQXUIDYRTHQTET-UHFFFAOYSA-N
Synonym 4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol
PubChem CID 3758882
IUPAC Name 4-amino-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])N

Specifications

Melting Point 152°C
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitrophenol
EINECSNumber (3)-0788
TSCA No