missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-nitrobenzophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 141°C |
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 242.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-3-nitrobenzophenone |
Safety and Handling
| TSCA | No |