accessibility menu, dialog, popup

UserName

NH2NOOO

CAS RN 31431-19-3

IUPAC Name: 4-benzoyl-2-nitroaniline
Synonyms: 4-benzoyl-2-nitroaniline
Molecular Weight (g/mol): 242.23
Molecular Formula: C13H10N2O3
InChi Key: NGOOFAMQPUEDJM-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1[N+]([O-])=O)C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results