missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxybenzoic acid, 98%

Catalog No. p-7080613
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1020706 5 g
AAA1020714 25 g
AAA1020722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1020706 Supplier Thermo Scientific Chemicals Supplier No. A1020706
Only null left

CAS: 2374-03-0 | C7H7NO3 | 153.137 g/mol

4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

Solubility
Soluble in chloroform, and aqeous ethanol, water (Partly miscible)

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2374-03-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00017094
InChI Key NFPYJDZQOKCYIE-UHFFFAOYSA-N
Synonym 3-hydroxy-4-aminobenzoic acid, 4-amino-3-hydroxy benzoic acid, benzoic acid, 4-amino-3-hydroxy, chembl1462, 2hdr, pubchem14418, 2-amino-5-carboxyphenol, intermediates-zcf02635, acmc-209g5k, 4-carboxy-2-hydroxyaniline
PubChem CID 137566
IUPAC Name 4-amino-3-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)N

Specifications

Melting Point ∼215°C (decomposition)
Quantity 5 g
Beilstein 509859
Solubility Information Soluble in chloroform,and aqeous ethanol,water (Partly miscible)
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-hydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only