accessibility menu, dialog, popup

UserName

CAS RN 2374-03-0

IUPAC Name: 4-amino-3-hydroxybenzoic acid
Synonyms: 3-hydroxy-4-aminobenzoic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: NFPYJDZQOKCYIE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1O)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (4)