missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chlorophenol hydrochloride, 98%

Catalog No. AAA1972006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1972006 5 g
AAA1972014 25 g
AAA1972022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1972006 Supplier Thermo Scientific Chemicals Supplier No. A1972006

CAS: 52671-64-4 | C6H7Cl2NO | 180.028 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52671-64-4
Molecular Formula C6H7Cl2NO
Molecular Weight (g/mol) 180.028
MDL Number MFCD00143110
InChI Key RFJVQGMBFQGZPV-UHFFFAOYSA-N
Synonym 4-amino-3-chlorophenol hydrochloride, 4-amino-3-chlorophenol hcl, 2-chloro-4-hydroxyaniline hydrochloride, phenol, 4-amino-3-chloro-, hydrochloride, 4-azanyl-3-chloranyl-phenol hydrochloride, c6h6clno.hcl, acmc-209l0t, ksc495i8f, 4-amino-3-chlorophenol, chloride, 3-chloro-4-aminophenol hydrochloride
PubChem CID 12598190
IUPAC Name 4-amino-3-chlorophenol;hydrochloride
SMILES C1=CC(=C(C=C1O)Cl)N.Cl

Specifications

Melting Point 254°C to 256°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 180.04
Percent Purity 98%
Chemical Name or Material 4-Amino-3-chlorophenol hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only