accessibility menu, dialog, popup

UserName

NH2OHCl

CAS RN 52671-64-4

IUPAC Name: 4-amino-3-chlorophenol
Synonyms: 4-amino-3-chlorophenol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: PNLPXABQLXSICH-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C=C1Cl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)