missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromobenzotrifluoride 98.0+%, TCI America™

Catalog No. A15781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15781G 1 g
A15785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15781G Supplier TCI America Supplier No. A15781G
Only null left

Chemical Identifiers

CAS 57946-63-1
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00042150
InChI Key QKRJIXSZTKOFTD-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl aniline, 4-amino-3-bromobenzotrifluoride, 2-bromo-4-trifluoromethyl-phenylamine, benzenamine, 2-bromo-4-trifluoromethyl, 3-bromo-4-aminobenzotrifluoride, 4-amino-3-bromo benzotrifluoride, buttpark 83\07-26, timtec-bb sbb000789, buttpark 44\03-51, 2-bromo-4-trifluoromethyl phenylamine
PubChem CID 93899
IUPAC Name 2-bromo-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Br)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 240.02
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4-Amino-3-bromobenzotrifluoride